siRNA Details
| Gene Symbol: Fam170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164485.1 | NP_001157957.1 | 2 | 894 | ||
| GenBank mRNA | AK076630.1 | - | 1 | 1670 | ||
| BC117714.1 | - | 1 | 784 | 40088057 | ||
| BC128503.1 | - | 1 | 784 | 40088058 | ||
| BC128504.1 | - | 1 | 784 | 40088059 | ||

Fam170b
Genomic Map