siRNA Details
| Gene Symbol: Chmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134078.4 | NP_598839.2 | 7, 8 | 1145 | ||
| GenBank mRNA | BB621791.1 | - | 3, 4 | 278 | ||
| BC024115.1 | - | 7, 8 | 1133 | 5100379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chmp7| Gene Symbol: Chmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134078.4 | NP_598839.2 | 7, 8 | 1145 | ||
| GenBank mRNA | BB621791.1 | - | 3, 4 | 278 | ||
| BC024115.1 | - | 7, 8 | 1133 | 5100379 | ||