siRNA Details
| Gene Symbol: Zfp957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033215.3 | NP_001028387.1 | 3 | 1893 | ||
| GenBank mRNA | AK139686.1 | - | 3 | 1823 | ||
| BC151187.1 | - | 2 | 1487 | 9088076 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp957| Gene Symbol: Zfp957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033215.3 | NP_001028387.1 | 3 | 1893 | ||
| GenBank mRNA | AK139686.1 | - | 3 | 1823 | ||
| BC151187.1 | - | 2 | 1487 | 9088076 | ||