siRNA Details
| Gene Symbol: Rcbtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170694.1 | NP_001164165.1 | 10 | 1097 | ||
| NM_134083.4 | NP_598844.3 | 10 | 1092 | |||
| GenBank mRNA | AK013540.1 | - | 1202 | |||
| AK031242.1 | - | 10 | 1058 | |||
| AK031248.1 | - | 10 | 1063 | |||
| AK147784.1 | - | 11 | 1098 | |||
| AK160881.1 | - | 11 | 1284 | |||
| BC003224.1 | - | 8 | 890 | 3482729 | ||
| JN086338.1 | - | 9 | 1013 | |||
| JN086339.1 | - | 10 | 1251 | |||

Rcbtb2
Genomic Map