siRNA Details
| Gene Symbol: Mtbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134092.3 | NP_598853.3 | 18 | 2399 | ||
| GenBank mRNA | AJ278508.1 | - | 18 | 2320 | ||
| AK080036.1 | - | 18 | 2213 | |||
| AK088652.1 | - | 8 | 1827 | |||
| AK133855.1 | - | 18 | 2247 | |||
| BC005500.1 | - | 11 | 1396 | |||
| BC052197.1 | - | 18 | 2399 | 6511752 | ||

Mtbp
Genomic Map