siRNA Details
| Gene Symbol: Dennd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081066.1 | NP_001074535.1 | 21 | 3608 | ||
| GenBank mRNA | AK053114.1 | - | 11 | 1942 | ||
| AK173046.1 | - | 21 | 3508 | |||
| BC030441.1 | - | 1 | 27 | |||
| BC132389.1 | - | 21 | 3496 | 40130666 | ||
| BC137791.1 | - | 21 | 3496 | 8860810 | ||
| BC144912.1 | - | 22 | 3624 | |||

Dennd3
Genomic Map