siRNA Details
| Gene Symbol: Card10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130859.2 | NP_570929.2 | 8 | 1438 | ||
| GenBank mRNA | AF363456.1 | - | 7 | 1287 | ||
| BC060203.1 | - | 8 | 1438 | 6855024 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Card10| Gene Symbol: Card10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130859.2 | NP_570929.2 | 8 | 1438 | ||
| GenBank mRNA | AF363456.1 | - | 7 | 1287 | ||
| BC060203.1 | - | 8 | 1438 | 6855024 | ||