siRNA Details
| Gene Symbol: 9030619P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136484.1 | - | 4 | 223 | ||
| BC087965.1 | - | 4 | 312 | |||
| BC147461.1 | - | 4 | 214 | 9056103 | ||
| BC147464.1 | - | 4 | 214 | 9056106 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9030619P08Rik| Gene Symbol: 9030619P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136484.1 | - | 4 | 223 | ||
| BC087965.1 | - | 4 | 312 | |||
| BC147461.1 | - | 4 | 214 | 9056103 | ||
| BC147464.1 | - | 4 | 214 | 9056106 | ||