siRNA Details
| Gene Symbol: Mfsd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134100.4 | NP_598861.1 | 2 | 381 | ||
| GenBank mRNA | AK087550.1 | - | 2 | 384 | ||
| AK089268.1 | - | 2 | 384 | |||
| AK165774.1 | - | 2 | 353 | |||
| AK197827.1 | - | 1 | 201 | |||
| BC009140.1 | - | 1 | 288 | 3598007 | ||

Mfsd5
Genomic Map