siRNA Details
| Gene Symbol: Cggbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178647.2 | NP_848762.1 | 3 | 822 | ||
| GenBank mRNA | AK049631.1 | - | 3 | 900 | ||
| AK052195.1 | - | 3 | 890 | |||
| AK083450.1 | - | 3 | 827 | |||
| AK086053.1 | - | 3 | 901 | |||
| BC031191.1 | - | 1 | 207 | |||
| BC043097.1 | - | 3 | 822 | 6405954 | ||
| BC057053.1 | - | 3 | 876 | 6827183 | ||

Cggbp1
Genomic Map