siRNA Details
| Gene Symbol: Ildr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285788.1 | NP_001272717.1 | 4 | 608 | ||
| NM_001285791.1 | NP_001272720.1 | 4 | 608 | |||
| NM_134109.2 | NP_598870.1 | 4 | 608 | |||
| GenBank mRNA | AK035504.1 | - | 4 | 537 | ||
| AK036844.1 | - | 4 | 534 | |||
| AK050806.1 | - | 4 | 554 | |||
| AK136284.1 | - | 4 | 608 | |||
| BC010485.1 | - | 4 | 548 | 3483341 | ||
| BC057644.1 | - | 4 | 605 | 4912789 | ||

Ildr1
Genomic Map