siRNA Details
| Gene Symbol: Eaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113401.1 | NP_001106872.1 | 5 | 684 | ||
| NM_001113405.1 | NP_001106876.1 | 3 | 326 | |||
| NM_134111.2 | NP_598872.1 | 5 | 674 | |||
| GenBank mRNA | AB081298.1 | - | 3 | 326 | ||
| AY034479.1 | - | 5 | 691 | |||
| AY049021.1 | - | 5 | 630 | |||
| BC004721.1 | - | 5 | 673 | 3582359 | ||
| BC056626.1 | - | 5 | 674 | 5008372 | ||

Eaf2
Genomic Map