siRNA Details
| Gene Symbol: Sft2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134114.2 | NP_598875.2 | 2 | 225 | ||
| GenBank mRNA | AK151338.1 | - | 2 | 156 | ||
| BC006777.1 | - | 2 | 180 | |||
| BC091770.1 | - | 2 | 226 | 4974098 | ||
| BC130232.1 | - | 2 | 149 | 40141936 | ||

Sft2d1
Genomic Map