siRNA Details
| Gene Symbol: Stk38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134115.2 | NP_598876.1 | 4 | 479 | ||
| GenBank mRNA | AK035731.1 | - | 4 | 488 | ||
| AK080179.1 | - | 4 | 488 | |||
| AY292399.1 | - | 4 | 343 | |||
| BC009658.1 | - | 4 | 479 | 3484764 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK153676.1 | - | 1420 | Chr. - - [-] on Build | ||

Stk38
Genomic Map