siRNA Details
| Gene Symbol: Tecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027179.1 | NP_081455.1 | 4, 5 | 366 | ||
| NM_134118.5 | NP_598879.1 | 5, 6 | 411 | |||
| GenBank mRNA | AK002384.1 | - | 5, 6 | 381 | ||
| AK010513.1 | - | 4, 5 | 302 | |||
| AK010984.1 | - | 5, 6 | 385 | |||
| AK013558.1 | - | 5, 6 | 367 | |||
| AK020708.1 | - | 4, 5 | 360 | |||
| BC019984.1 | - | 5, 6 | 397 | 4240045 | ||
| BC094046.1 | - | 5, 6 | 511 | 5064002 | ||
| BY278351.1 | - | 4, 5 | 366 | |||

Tecr
Genomic Map