siRNA Details
| Gene Symbol: Dlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286013.1 | NP_001272942.1 | 5 | 490 | ||
| NM_001286028.1 | NP_001272957.1 | 5 | 377 | |||
| GenBank mRNA | BC118057.1 | - | 5 | 377 | 40105170 | |
| BC122518.1 | - | 6 | 405 | 40105175 | ||
| BG868693.1 | - | 5 | 470 | |||
| FM180474.1 | - | 5 | 490 | |||

Dlk2
Genomic Map