siRNA Details
| Gene Symbol: Rab31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133685.2 | NP_598446.2 | 6 | 621 | ||
| GenBank mRNA | AB232617.1 | - | 6 | 413 | ||
| AK007048.1 | - | 2 | 362 | |||
| AK016608.1 | - | 2 | 351 | |||
| AK051270.1 | - | 6 | 541 | |||
| AK156677.1 | - | 6 | 622 | |||
| AK159095.1 | - | 6 | 510 | |||
| BC013063.1 | - | 6 | 550 | 3497938 | ||

Rab31
Genomic Map