siRNA Details
| Gene Symbol: Ift140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134126.3 | NP_598887.3 | 6, 7 | 1654 | ||
| GenBank mRNA | AK129167.1 | - | 3, 4 | 345 | ||
| AK149331.1 | - | 6, 7 | 883 | |||
| AK163782.1 | - | 6, 7 | 1282 | |||
| BC139005.1 | - | 6, 7 | 883 | 8862027 | ||
| BC145494.1 | - | 6, 7 | 883 | |||
| DQ266093.1 | - | 6, 7 | 1017 | |||

Ift140
Genomic Map