siRNA Details
| Gene Symbol: Ift140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134126.3 | NP_598887.3 | 9 | 1898 | ||
| GenBank mRNA | AK129167.1 | - | 6 | 589 | ||
| AK149331.1 | - | 9 | 1127 | |||
| AK163782.1 | - | 9 | 1526 | |||
| BC139005.1 | - | 9 | 1127 | 8862027 | ||
| BC145494.1 | - | 9 | 1127 | |||
| DQ266093.1 | - | 9 | 1261 | |||

Ift140
Genomic Map