siRNA Details
| Gene Symbol: AI413582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002895.2 | NP_001002895.2 | 4 | 316 | ||
| GenBank mRNA | AK131593.1 | - | 4 | 311 | ||
| AK131681.1 | - | 4 | 291 | |||
| BC028464.1 | - | 4 | 289 | 1264774 | ||
| BY244090.1 | - | 3 | 468 | |||

AI413582
Genomic Map