siRNA Details
| Gene Symbol: Dhx57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163759.1 | NP_001157231.1 | 21 | 4098 | ||
| NM_198942.2 | NP_945180.2 | 20 | 3939 | |||
| GenBank mRNA | AK035344.1 | - | 5 | 780 | ||
| AK162192.1 | - | 3 | 342 | |||
| BC026474.1 | - | 8 | 1080 | |||
| BC037692.1 | - | 9 | 1175 | |||
| BC062952.1 | - | 20 | 3939 | 6853031 | ||
| BC065169.1 | - | 21 | 3765 | 6831314 | ||
| BC066091.1 | - | 20 | 3601 | 6841207 | ||
| BC079618.1 | - | 20 | 4219 | |||

Dhx57
Genomic Map