siRNA Details
| Gene Symbol: Dhx57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163759.1 | NP_001157231.1 | 5 | 1313 | ||
| NM_198942.2 | NP_945180.2 | 4 | 1154 | |||
| GenBank mRNA | AK053628.1 | - | 5 | 981 | ||
| BC062952.1 | - | 4 | 1154 | 6853031 | ||
| BC065169.1 | - | 5 | 980 | 6831314 | ||
| BC066091.1 | - | 4 | 816 | 6841207 | ||

Dhx57
Genomic Map