siRNA Details
| Gene Symbol: Oard1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289490.1 | NP_001276419.1 | 5, 6 | 776 | ||
| NM_001289491.1 | NP_001276420.1 | 5, 6 | 501 | |||
| NM_207219.4 | NP_997102.2 | 5, 6 | 641 | |||
| GenBank mRNA | AK148136.1 | - | 5, 6 | 763 | ||
| AK153512.1 | - | 5, 6 | 564 | |||
| BC022574.1 | - | 4, 5 | 420 | 3989373 | ||
| BC157987.1 | - | 5, 6 | 526 | 9007303 | ||

Oard1
Genomic Map