siRNA Details
| Gene Symbol: Smim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134133.2 | NP_598894.1 | 2 | 1532 | ||
| GenBank mRNA | AF313412.1 | - | 2 | 1074 | ||
| AK143383.1 | - | 2 | 1536 | |||
| AK155873.1 | - | 2 | 1178 | |||
| AK168283.1 | - | 2 | 1355 | |||
| AK171810.1 | - | 2 | 1190 | |||
| BC012898.1 | - | 2 | 1040 | 4013357 | ||

Smim3
Genomic Map