siRNA Details
| Gene Symbol: Gramd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026240.2 | NP_080516.2 | 13 | 1386 | ||
| GenBank mRNA | AK018551.1 | - | 14 | 1636 | ||
| AK018691.1 | - | 13 | 1372 | |||
| AK161329.1 | - | 13 | 1319 | |||
| AK164050.1 | - | 13 | 1371 | |||
| BC057978.1 | - | 13 | 1361 | 5044170 | ||

Gramd3
Genomic Map