siRNA Details
| Gene Symbol: Rrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199447.2 | NP_955518.1 | 29 | 3566 | ||
| GenBank mRNA | AK122346.1 | - | 28 | 4680 | ||
| AK146550.1 | - | 29 | 3574 | |||
| BC056232.1 | - | 29 | 3598 | 30060906 | ||
| BC062977.1 | - | 29 | 3566 | 6853905 | ||

Rrp12
Genomic Map