siRNA Details
| Gene Symbol: Cyp2c50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167875.1 | NP_001161347.1 | 5 | 679 | ||
| NM_134144.2 | NP_598905.2 | 6 | 856 | |||
| GenBank mRNA | AY206873.1 | - | 6 | 835 | ||
| BC011222.1 | - | 6 | 839 | 4195223 | ||
| BC051050.1 | - | 5 | 670 | 5052536 | ||

Cyp2c50
Genomic Map