siRNA Details
| Gene Symbol: Kazald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178929.4 | NP_849260.2 | 3 | 690 | ||
| GenBank mRNA | AK030678.1 | - | 3 | 689 | ||
| AK155095.1 | - | 4 | 935 | |||
| BC115642.1 | - | 3 | 681 | 40103018 | ||
| BC115643.1 | - | 3 | 678 | |||

Kazald1
Genomic Map