siRNA Details
| Gene Symbol: Otub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134150.2 | NP_598911.1 | 6 | 623 | ||
| GenBank mRNA | AK027996.1 | - | 6 | 610 | ||
| AK087195.1 | - | 6 | 610 | |||
| AK140070.1 | - | 6 | 610 | |||
| AK145273.1 | - | 6 | 613 | |||
| AK145970.1 | - | 6 | 615 | |||
| AK191684.1 | - | 1 | 177 | |||
| BC022575.1 | - | 6 | 623 | 5005025 | ||
| BC054410.1 | - | 6 | 591 | |||

Otub1
Genomic Map