siRNA Details
| Gene Symbol: Yars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134151.4 | NP_598912.4 | 10 | 1333 | ||
| GenBank mRNA | AK028785.1 | - | 10 | 1195 | ||
| AK043837.1 | - | 10 | 1257 | |||
| AK076634.1 | - | 10 | 1178 | |||
| AK078313.1 | - | 2 | 1911 | |||
| AK139579.1 | - | 10 | 1178 | |||
| AK145356.1 | - | 10 | 1226 | |||
| AK151880.1 | - | 10 | 1185 | |||
| AK169708.1 | - | 10 | 1109 | |||
| BC013552.1 | - | 10 | 1264 | 4236514 | ||
| BC022143.1 | - | 10 | 1228 | 5101324 | ||
| BC026615.1 | - | 10 | 1240 | 4976999 | ||

Yars
Genomic Map