siRNA Details
| Gene Symbol: Yars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134151.4 | NP_598912.4 | 8 | 1090 | ||
| GenBank mRNA | AK028785.1 | - | 8 | 952 | ||
| AK043837.1 | - | 8 | 1014 | |||
| AK076634.1 | - | 8 | 935 | |||
| AK139579.1 | - | 8 | 935 | |||
| AK145356.1 | - | 8 | 983 | |||
| AK151880.1 | - | 8 | 942 | |||
| AK169708.1 | - | 8 | 866 | |||
| AK219154.1 | - | 2 | 114 | |||
| BC013552.1 | - | 8 | 1021 | 4236514 | ||
| BC022143.1 | - | 8 | 985 | 5101324 | ||
| BC026615.1 | - | 8 | 997 | 4976999 | ||

Yars
Genomic Map