siRNA Details
| Gene Symbol: Vps37c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181403.2 | NP_852068.1 | 5 | 1079 | ||
| GenBank mRNA | AK030717.1 | - | 4 | 950 | ||
| AK150606.1 | - | 5 | 1060 | |||
| AK158833.1 | - | 4 | 818 | |||
| AK159309.1 | - | 5 | 1152 | |||
| AK165826.1 | - | 5 | 1147 | |||
| BC025865.1 | - | 5 | 1079 | 5029143 | ||

Vps37c
Genomic Map