siRNA Details
| Gene Symbol: Gbf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178930.3 | NP_849261.2 | 33 | 4558 | ||
| GenBank mRNA | AK172919.1 | - | 30 | 4151 | ||
| BC023791.1 | - | 7 | 976 | |||
| BC024129.1 | - | 16 | 2216 | |||
| BC026621.1 | - | 4 | 590 | |||
| BC034745.1 | - | 18 | 2323 | |||
| BC059883.1 | - | 10 | 1223 | |||
| BC060728.1 | - | 25 | 3978 | |||
| BC076569.1 | - | 33 | 4558 | 6409170 | ||
| BC082336.1 | - | 34 | 4553 | |||

Gbf1
Genomic Map