siRNA Details
| Gene Symbol: AW112010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA086638.1 | - | 3 | 326 | ||
| AA467412.1 | - | 3 | 261 | |||
| AK153119.1 | - | 3 | 174 | |||
| EF660528.1 | - | 3 | 278 | |||
| EH107460.1 | - | 3 | 211 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AW112010| Gene Symbol: AW112010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA086638.1 | - | 3 | 326 | ||
| AA467412.1 | - | 3 | 261 | |||
| AK153119.1 | - | 3 | 174 | |||
| EF660528.1 | - | 3 | 278 | |||
| EH107460.1 | - | 3 | 211 | |||