siRNA Details
| Gene Symbol: AW112010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK153119.1 | - | 3 | 413 | ||
| BG091196.1 | - | 1 | 160 | |||
| EF660528.1 | - | 3 | 517 | |||
| EH107460.1 | - | 3 | 450 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AW112010| Gene Symbol: AW112010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK153119.1 | - | 3 | 413 | ||
| BG091196.1 | - | 1 | 160 | |||
| EF660528.1 | - | 3 | 517 | |||
| EH107460.1 | - | 3 | 450 | |||