siRNA Details
| Gene Symbol: OR1F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012360.1 | NP_036492.1 | 1 | 322 | ||
| XM_011522506.2 | XP_011520808.1 | 4 | 769 | |||
| XM_011522507.2 | XP_011520809.1 | 2 | 1192 | |||
| GenBank mRNA | BC069335.1 | - | 1 | 322 | 7262265 | |
| Y14442.1 | - | 1 | 322 | |||

OR1F1
Genomic Map