siRNA Details
| Gene Symbol: Pdzd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033222.3 | NP_001028394.2 | 3, 4 | 1280 | ||
| GenBank mRNA | AK041847.1 | - | 3, 4 | 734 | ||
| AK145020.1 | - | 3, 4 | 1280 | |||
| AK195576.1 | - | 1, 2 | 65 | |||
| BC141370.1 | - | 3, 4 | 1185 | 9055983 | ||
| BC141371.1 | - | 3, 4 | 1185 | 9055984 | ||

Pdzd8
Genomic Map