siRNA Details
| Gene Symbol: OR2C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012368.2 | NP_036500.2 | 1 | 934 | ||
| GenBank mRNA | BC069158.1 | - | 1 | 882 | 7217019 | |
| BC069456.1 | - | 1 | 882 | 7262277 | ||
| BC126269.1 | - | 1 | 934 | 8991985 | ||
| BC130328.1 | - | 1 | 934 | 40146359 | ||

OR2C1
Genomic Map