siRNA Details
| Gene Symbol: Unc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153131.3 | NP_694771.1 | 2 | 490 | ||
| GenBank mRNA | AK122575.1 | - | 2 | 258 | ||
| BC058084.1 | - | 2 | 511 | 6813463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc5a| Gene Symbol: Unc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153131.3 | NP_694771.1 | 2 | 490 | ||
| GenBank mRNA | AK122575.1 | - | 2 | 258 | ||
| BC058084.1 | - | 2 | 511 | 6813463 | ||