siRNA Details
| Gene Symbol: Mkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181860.1 | NP_862908.1 | 3, 4 | 328 | ||
| GenBank mRNA | BC028459.1 | - | 3, 4 | 328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mkl2| Gene Symbol: Mkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181860.1 | NP_862908.1 | 3, 4 | 328 | ||
| GenBank mRNA | BC028459.1 | - | 3, 4 | 328 | ||