siRNA Details
Gene Symbol: Lgr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172671.2 | NP_766259.2 | 18 | 2377 | ||
GenBank mRNA | AK033699.1 | - | 3 | 497 | ||
AK044357.1 | - | 17 | 1685 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lgr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172671.2 | NP_766259.2 | 18 | 2377 | ||
GenBank mRNA | AK033699.1 | - | 3 | 497 | ||
AK044357.1 | - | 17 | 1685 |