siRNA Details
| Gene Symbol: Gimap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243199.1 | NP_001230128.1 | 4 | 873 | ||
| NM_001243200.1 | NP_001230129.1 | 4 | 938 | |||
| NM_001243201.1 | NP_001230130.1 | 4 | 414 | |||
| NM_174990.4 | NP_778155.2 | 3 | 1322 | |||
| NM_175048.3 | NP_778213.2 | 5 | 475 | |||
| GenBank mRNA | AA795946.1 | - | 5 | 450 | ||
| AK088228.1 | - | 5 | 593 | |||
| BC005577.1 | - | 4 | 863 | 3968418 | ||
| BC098226.1 | - | 3 | 387 | |||

Gimap4
Genomic Map