siRNA Details
| Gene Symbol: Gimap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243199.1 | NP_001230128.1 | 3 | 666 | ||
| NM_001243200.1 | NP_001230129.1 | 4 | 282 | |||
| NM_174990.4 | NP_778155.2 | 3 | 666 | |||
| NM_175048.3 | NP_778213.2 | 4 | 282 | |||
| GenBank mRNA | AA543119.1 | - | 4 | 256 | ||
| AA795946.1 | - | 4 | 256 | |||
| AK088228.1 | - | 4 | 400 | |||
| BC005577.1 | - | 3 | 656 | 3968418 | ||
| CA575154.1 | - | 1 | 34 | |||
| CA576106.1 | - | 1 | 338 | |||
| CB232465.1 | - | 4 | 313 | |||

Gimap4
Genomic Map