siRNA Details
| Gene Symbol: Wwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276292.1 | NP_001263221.1 | 19 | 2348 | ||
| NM_177327.5 | NP_796301.2 | 20 | 2510 | |||
| GenBank mRNA | AK031762.1 | - | 4 | 443 | ||
| AK032232.1 | - | 19 | 2176 | |||
| AK033138.1 | - | 12 | 1324 | |||
| AK137588.1 | - | 20 | 2435 | |||
| AK168567.1 | - | 20 | 2374 | |||
| BC021470.1 | - | 3 | 325 | |||
| BC026829.1 | - | 19 | 2286 | 4209397 | ||
| BC055937.1 | - | 2 | 229 | |||
| BC150812.1 | - | 19 | 2345 | 9087723 | ||

Wwp1
Genomic Map