siRNA Details
| Gene Symbol: Ovol2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026924.3 | NP_081200.2 | 4 | 940 | ||
| NM_152947.2 | NP_694455.2 | 4 | 681 | |||
| GenBank mRNA | AB101294.1 | - | 4 | 843 | ||
| AB101295.1 | - | 3 | 885 | |||
| AB101296.1 | - | 4 | 1015 | |||
| AK007360.1 | - | 3 | 566 | |||
| AK167697.1 | - | 4 | 931 | |||
| AY090537.1 | - | 4 | 751 | |||
| AY090538.1 | - | 4 | 681 | |||
| BC031771.1 | - | 4 | 940 | 4219296 | ||
| BC057210.1 | - | 4 | 961 | 5329625 | ||
| BC094445.1 | - | 4 | 952 | 4981833 | ||

Ovol2
Genomic Map