siRNA Details
| Gene Symbol: Rnh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172100.1 | NP_001165571.1 | 3 | 370 | ||
| NM_001172101.1 | NP_001165572.1 | 3 | 373 | |||
| NM_145135.3 | NP_660117.2 | 3 | 521 | |||
| GenBank mRNA | AF071546.1 | - | 2 | 241 | ||
| AK133822.1 | - | 3 | 301 | |||
| AK141351.1 | - | 3 | 346 | |||
| AK145173.1 | - | 3 | 355 | |||
| AK147059.1 | - | 3 | 307 | |||
| AK188203.1 | - | 128 | ||||
| BC010331.1 | - | 3 | 390 | 2654341 | ||
| BM946973.1 | - | 3 | 521 | |||

Rnh1
Genomic Map