siRNA Details
| Gene Symbol: Slc12a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133648.2 | NP_598409.2 | 23 | 3062 | ||
| NM_133649.2 | NP_598410.2 | 23 | 3723 | |||
| GenBank mRNA | AF211854.1 | - | 23 | 3218 | ||
| AF211855.1 | - | 23 | 3062 | |||
| AK133006.1 | - | 23 | 4213 | |||
| BC036323.1 | - | 10 | 1295 | |||
| BC051061.1 | - | 2 | 1570 | |||
| BC062099.1 | - | 3678 | 6844732 | |||

Slc12a6
Genomic Map