siRNA Details
| Gene Symbol: Slc12a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133648.2 | NP_598409.2 | 16 | 1997 | ||
| NM_133649.2 | NP_598410.2 | 16 | 2658 | |||
| GenBank mRNA | AF211854.1 | - | 16 | 2153 | ||
| AF211855.1 | - | 16 | 1997 | |||
| AK133006.1 | - | 16 | 3148 | |||
| AK137387.1 | - | 15 | 1977 | |||
| BC036323.1 | - | 3 | 230 | |||
| BC062099.1 | - | 2613 | 6844732 | |||

Slc12a6
Genomic Map