siRNA Details
| Gene Symbol: Tm6sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291282.1 | NP_001278211.1 | 5, 6 | 429 | ||
| NM_145375.4 | NP_663350.2 | 5, 6 | 937 | |||
| GenBank mRNA | AK046714.1 | - | 5, 6 | 838 | ||
| AK083132.1 | - | 5, 6 | 940 | |||
| AK135100.1 | - | 5, 6 | 919 | |||
| AK136581.1 | - | 5, 6 | 430 | |||
| AK163621.1 | - | 5, 6 | 862 | |||
| AK172109.1 | - | 5, 6 | 853 | |||
| AK172145.1 | - | 5, 6 | 838 | |||
| AY512939.1 | - | 1, 2 | 1054 | |||
| BC023123.1 | - | 5, 6 | 819 | 3995280 | ||

Tm6sf1
Genomic Map