siRNA Details
| Gene Symbol: Adgrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174991.3 | NP_778156.2 | 7 | 2304 | ||
| GenBank mRNA | AK147456.1 | - | 7 | 2289 | ||
| AK147459.1 | - | 7 | 2257 | |||
| AK147494.1 | - | 7 | 2091 | |||
| AK147600.1 | - | 7 | 2066 | |||
| AK147607.1 | - | 7 | 2308 | |||
| AK220481.1 | - | 7 | 1683 | |||
| AY168408.1 | - | 7 | 1816 | |||
More Information
Set Membership: |
|

Adgrb1
Genomic Map